Gene Transcripts

Gene Symbol
Cldnd2
Entrez Gene ID
499140
Gene Name
claudin domain containing 2
Gene Aliases
RGD1566413
Chromosome Location
Chr. 1: 98487631 - 98490315 on Build Rnor_6.0
UniGene ID
Rn.125310
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109143.1 NP_001102613.1 4 528

View Full Product Details