Gene Transcripts

Gene Symbol
Kctd21
Entrez Gene ID
499209
Gene Name
potassium channel tetramerization domain containing 21
Gene Aliases
RGD1559529
Chromosome Location
Chr. 1: 162307775 - 162325269 on Build Rnor_6.0
UniGene ID
Rn.214892
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109151.1 NP_001102621.1 1 362
XM_006229781.2 XP_006229843.1 2 433

View Full Product Details