Gene Transcripts

Gene Symbol
Gsg1l
Entrez Gene ID
499263
Gene Name
GSG1-like
Gene Aliases
RGD1562278
Chromosome Location
Chr. 1: 197271704 - 197482622 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759923.1 XP_008758145.1 1 374

View Full Product Details