Gene Transcripts

Gene Symbol
Tti1
Entrez Gene ID
499935
Gene Name
TELO2 interacting protein 1
Gene Aliases
RGD1562582
Chromosome Location
Chr. 3: 154463108 - 154506950 on Build Rnor_6.0
UniGene ID
Rn.48856
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134619.1 NP_001128091.1 1 1607
XM_006235428.2 XP_006235490.1 2 1684
XM_008762375.1 XP_008760597.1 2 2447

View Full Product Details