Gene Transcripts

Gene Symbol
Apcdd1l
Entrez Gene ID
499949
Gene Name
adenomatosis polyposis coli down-regulated 1-like
Gene Aliases
Chromosome Location
Chr. 3: 171883156 - 171958156 on Build Rnor_6.0
UniGene ID
Rn.161875
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235711.2 XP_006235773.1 3 816
XM_008762580.1 XP_008760802.1 3 2898
GenBank mRNA BC081904.1 - 656

View Full Product Details