Gene Transcripts

Gene Symbol
Klhl42
Entrez Gene ID
500367
Gene Name
kelch-like family, member 42
Gene Aliases
Klhdc5, RGD1560924
Chromosome Location
Chr. 4: 181481147 - 181502916 on Build Rnor_6.0
UniGene ID
Rn.203842
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109257.1 NP_001102727.1 1, 2 884

View Full Product Details