Gene Transcripts

Gene Symbol
Adam4l1
Entrez Gene ID
500688
Gene Name
a disintegrin and metallopeptidase domain 4-like 1
Gene Aliases
Chromosome Location
Chr. 6: 105007053 - 105009516 on Build Rnor_6.0
UniGene ID
Rn.224743
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109277.1 NP_001102747.1 1 2016
GenBank mRNA BC079009.1 - 1 1994

View Full Product Details