Gene Transcripts

Gene Symbol
Tmem89
Entrez Gene ID
501056
Gene Name
transmembrane protein 89
Gene Aliases
RGD1564575
Chromosome Location
Chr. 8: 117660975 - 117661869 on Build Rnor_6.0
UniGene ID
Rn.202006
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163477.1 NP_001156949.1 1 237

View Full Product Details