Gene Transcripts

Gene Symbol
Lrrc73
Entrez Gene ID
501101
Gene Name
leucine rich repeat containing 73
Gene Aliases
nod3l
Chromosome Location
Chr. 9: 17111176 - 17115276 on Build Rnor_6.0
UniGene ID
Rn.51041
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024360.2 NP_001019531.1 1 277
XM_008766878.1 XP_008765100.1 1 1376
GenBank mRNA AB210855.1 - 1 177
BC128712.1 - 1 277 7133504

View Full Product Details