Gene Transcripts

Gene Symbol
Krtap14
Entrez Gene ID
501768
Gene Name
keratin associated protein 14
Gene Aliases
RGD1564302
Chromosome Location
Chr. 11: 28776761 - 28777603 on Build Rnor_6.0
UniGene ID
Rn.218670
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109326.1 NP_001102796.1 1 487

View Full Product Details