Gene Transcripts

Gene Symbol
Lor
Entrez Gene ID
502541
Gene Name
loricrin
Gene Aliases
RGD1559993
Chromosome Location
Chr. 2: 191984455 - 191989058 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761253.1 XP_008759475.1 2 1868

View Full Product Details