Gene Transcripts

Gene Symbol
Odam
Entrez Gene ID
641555
Gene Name
odontogenic, ameloblast asssociated
Gene Aliases
Apin
Chromosome Location
Chr. 14: 21842990 - 21852216 on Build Rnor_6.0
UniGene ID
Rn.65661
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044274.1 NP_001037739.1 10 743
GenBank mRNA DQ198380.1 - 10 743

View Full Product Details