Gene Transcripts

Gene Symbol
Krt83
Entrez Gene ID
681126
Gene Name
keratin 83, type II
Gene Aliases
Chromosome Location
Chr. 7: 143078996 - 143085833 on Build Rnor_6.0
UniGene ID
Rn.217931
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101675.1 NP_001095145.1 4 697
GenBank mRNA AB354637.1 - 4 697

View Full Product Details