Gene Transcripts

Gene Symbol
Ppp2r3b
Entrez Gene ID
682033
Gene Name
protein phosphatase 2, regulatory subunit B'', beta
Gene Aliases
Chromosome Location
Chr. 14: 1469748 - 1476353 on Build Rnor_6.0
UniGene ID
Rn.233844
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139492.1 NP_001132964.1 6 1262
GenBank mRNA BC161890.1 - 6 1262

View Full Product Details