Gene Transcripts

Gene Symbol
Isx
Entrez Gene ID
682968
Gene Name
intestine-specific homeobox
Gene Aliases
Chromosome Location
Chr. 19: 14198899 - 14219269 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751833.3 XP_003751881.1 1 170
XM_008772332.1 XP_008770554.1 1 170

View Full Product Details