Gene Transcripts

Gene Symbol
Slc2a7
Entrez Gene ID
100362644
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 7
Gene Aliases
Chromosome Location
Chr. 5: 167188086 - 167211608 on Build Rnor_6.0
UniGene ID
Rn.217486
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101020.1 NP_001094490.1 434
GenBank mRNA EF501821.1 - 434

View Full Product Details