Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP6AP2
Entrez Gene ID
705048
Gene Name
ATPase H+ transporting accessory protein 2
Gene Aliases
EGK_20395
Chromosome Location
Chr.X: 40420640 - 40447093 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257625.1 NP_001244554.1 - 266 61
GenBank mRNA JU332431.1 - - 266 61
JU473893.1 - - 192 61
JV045853.1 - - 218 61

View Full Product Details