Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP131
Entrez Gene ID
718613
Gene Name
centrosomal protein 131
Gene Aliases
AZI1
Chromosome Location
Chr.16: 77525764 - 77563623 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028836908.1 XP_028692741.1 - 885 61
XM_028836909.1 XP_028692742.1 - 882 61
XM_028836910.1 XP_028692743.1 - 886 61
XM_028836912.1 XP_028692745.1 - 885 61
XM_028836913.1 XP_028692746.1 - 886 61
XM_028836914.1 XP_028692747.1 - 885 61
XM_028836915.1 XP_028692748.1 - 886 61
GenBank mRNA JU474774.1 - - 793 61
JU474775.1 - - 793 61
JV609571.1 - - 791 61

View Full Product Details