Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIT1
Entrez Gene ID
717985
Gene Name
Ras like without CAAX 1
Gene Aliases
EGK_01400
Chromosome Location
Chr.1: 95084097 - 95097525 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261801.1 NP_001248730.1 - 569 64
XM_015110011.2 XP_014965497.1 - 684 64
XM_028845924.1 XP_028701757.1 - 695 64
XM_028845937.1 XP_028701770.1 - 613 64
GenBank mRNA JU335790.1 - - 471 64
JU472493.1 - - 586 64
JV044219.1 - - 569 64

View Full Product Details