Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TXNDC17
Entrez Gene ID
712910
Gene Name
thioredoxin domain containing 17
Gene Aliases
EGK_08088
Chromosome Location
Chr.16: 6404236 - 6406274 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266428.1 NP_001253357.1 - 315 81
GenBank mRNA JU321845.1 - - 380 81
JU477540.1 - - 315 81
JV048961.1 - - 387 81

View Full Product Details