Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDRGK1
Entrez Gene ID
717226
Gene Name
DDRGK domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 63300533 - 63312999 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194674.3 NP_001181603.2 - 804 74
GenBank mRNA JU323166.1 - - 797 74
JV659436.1 - - 788 74

View Full Product Details