Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNRPD1
Entrez Gene ID
698965
Gene Name
small nuclear ribonucleoprotein D1 polypeptide
Gene Aliases
-
Chromosome Location
Chr.18: 60785421 - 60801906 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260829.1 NP_001247758.1 - 122 83
GenBank mRNA JU336042.1 - - 136 83
JU476907.1 - - 60 83
JV048017.1 - - 122 83
JV517890.1 - - 106 83
JV555563.1 - - 141 83

View Full Product Details