Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APLN
Entrez Gene ID
702695
Gene Name
apelin
Gene Aliases
-
Chromosome Location
Chr.X: 125900749 - 125910531 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015128162.2 XP_014983648.1 - 657 102
GenBank mRNA JU336931.1 - - 511 102

View Full Product Details