Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A14
Entrez Gene ID
710743
Gene Name
solute carrier family 16 member 14
Gene Aliases
EGK_04873
Chromosome Location
Chr.12: 117466790 - 117504478 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015111302.2 XP_014966788.1 - 732 87
XM_028831195.1 XP_028687028.1 - 733 87
GenBank mRNA JU326336.1 - - 253 87
JU326337.1 - - 253 87

View Full Product Details