Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD10
Entrez Gene ID
707443
Gene Name
potassium channel tetramerization domain containing 10
Gene Aliases
-
Chromosome Location
Chr.11: 109093895 - 109123004 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261187.1 NP_001248116.1 - 437 76
XM_028828895.1 XP_028684728.1 - 478 76
GenBank mRNA JU336639.1 - - 455 76
JU477440.1 - - 446 76
JV048403.1 - - 437 76

View Full Product Details