Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP69
Entrez Gene ID
704003
Gene Name
cilia and flagella associated protein 69
Gene Aliases
-
Chromosome Location
Chr.3: 115758959 - 115826978 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015134138.2 XP_014989624.1 - 1782 143
XM_015134139.2 XP_014989625.1 - 1783 143
XM_015134140.2 XP_014989626.1 - 1324 143
XM_028846088.1 XP_028701921.1 - 1728 143
XM_028846089.1 XP_028701922.1 - 1783 143

View Full Product Details