Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CETN3
Entrez Gene ID
695223
Gene Name
centrin 3
Gene Aliases
centrin-3
Chromosome Location
Chr.6: 87258052 - 87273731 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266092.2 NP_001253021.1 - 272 128
GenBank mRNA JU332582.1 - - 263 128
JU474049.1 - - 265 128
JV045969.1 - - 275 128
JV640527.1 - - 272 128

View Full Product Details