Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL20
Entrez Gene ID
694689
Gene Name
interleukin 20
Gene Aliases
-
Chromosome Location
Chr.1: 64378251 - 64384912 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015117964.2 XP_014973450.1 - 542 108

View Full Product Details