Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM17
Entrez Gene ID
694384
Gene Name
transmembrane protein 17
Gene Aliases
EGK_05371
Chromosome Location
Chr.13: 46252346 - 46259161 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015112568.2 XP_014968054.1 - 615 113
GenBank mRNA JU333302.1 - - 222 113
JU333303.1 - - 222 113
JU333304.1 - - 222 113
JV703741.1 - - 416 113

View Full Product Details