Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN1
Entrez Gene ID
704330
Gene Name
claudin 1
Gene Aliases
EGK_12212, claudin-1
Chromosome Location
Chr.2: 7536511 - 7551873 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193969.1 NP_001180898.1 - 497 71
GenBank mRNA JV045368.1 - - 248 71
JV635805.1 - - 471 71

View Full Product Details