Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXF1
Entrez Gene ID
694454
Gene Name
forkhead box F1
Gene Aliases
-
Chromosome Location
Chr.20: 73524125 - 73528071 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001084773.4 XP_001084773.2 - 1029 66

View Full Product Details