Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF41
Entrez Gene ID
711969
Gene Name
ring finger protein 41
Gene Aliases
EGK_03792
Chromosome Location
Chr.11: 55765562 - 55797777 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194648.2 NP_001181577.1 - 603 60
XM_015151956.2 XP_015007442.1 - 760 60
XM_015151957.2 XP_015007443.1 - 860 60
XM_015151960.2 XP_015007446.1 - 809 60
XM_015151961.2 XP_015007447.1 - 628 60
XM_028828955.1 XP_028684788.1 - 876 60
XM_028828956.1 XP_028684789.1 - 546 60
GenBank mRNA JU332442.1 - - 730 60
JU473908.1 - - 651 60
JV045863.1 - - 718 60
JV601081.1 - - 725 60

View Full Product Details