Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL7A
Entrez Gene ID
693894
Gene Name
methyltransferase like 7A
Gene Aliases
EGK_03645
Chromosome Location
Chr.11: 50547130 - 50556503 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193711.2 NP_001180640.2 - 582 63
XM_028828802.1 XP_028684635.1 - 927 63
XM_028828803.1 XP_028684636.1 - 1489 63
GenBank mRNA JV647601.1 - - 811 63

View Full Product Details