Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUS2
Entrez Gene ID
702051
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, EGK_12919
Chromosome Location
Chr.20: 55234301 - 55293070 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015126450.2 XP_014981936.1 - 985 84
XM_015126451.2 XP_014981937.1 - 833 84
XM_028841542.1 XP_028697375.1 - 758 84
XM_028841543.1 XP_028697376.1 - 628 84
XM_028841544.1 XP_028697377.1 - 642 84
GenBank mRNA JV064396.1 - - 576 84
JV472713.1 - - 568 84

View Full Product Details