Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NME8
Entrez Gene ID
705583
Gene Name
NME/NM23 family member 8
Gene Aliases
TXNDC3
Chromosome Location
Chr.3: 77274132 - 77327535 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015133693.2 XP_014989179.2 - 1757 115
XM_015133694.2 XP_014989180.2 - 1281 115
XM_015133695.2 XP_014989181.2 - 1664 115

View Full Product Details