Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH17
Entrez Gene ID
695637
Gene Name
protocadherin 17
Gene Aliases
EGK_09351
Chromosome Location
Chr.17: 36263150 - 36361516 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265711.1 NP_001252640.1 - 3521 58
XM_015121142.2 XP_014976628.1 - 6298 58
XM_015121143.2 XP_014976629.1 - 6193 58
XM_028837095.1 XP_028692928.1 - 6088 58
GenBank mRNA JV565011.1 - - 3521 58
JV712201.1 - - 3522 58

View Full Product Details