Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP9
Entrez Gene ID
693821
Gene Name
THAP domain containing 9
Gene Aliases
EGK_15718
Chromosome Location
Chr.5: 50769338 - 50789693 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086766.4 XP_001086766.1 - 223 83
XM_015138416.2 XP_014993902.1 - 223 83
XM_015138417.2 XP_014993903.1 - 102 83

View Full Product Details