Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLCO1B3
Entrez Gene ID
574354
Gene Name
solute carrier organic anion transporter family member 1B3
Gene Aliases
-
Chromosome Location
Chr.11: 22235443 - 22327301 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032941.1 NP_001028113.1 - 1159 78
XM_015151097.2 XP_015006583.1 - 1301 78
XM_028828782.1 XP_028684615.1 - 1273 78
GenBank mRNA AY787037.1 - - 1159 78

View Full Product Details