Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXCL13
Entrez Gene ID
574224
Gene Name
C-X-C motif chemokine ligand 13
Gene Aliases
BLC, EGK_15756
Chromosome Location
Chr.5: 56100955 - 56105858 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032880.1 NP_001028052.1 - 62 75
GenBank mRNA AF449284.1 - - 62 75

View Full Product Details