Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CGB8
Entrez Gene ID
574324
Gene Name
chorionic gonadotropin, beta polypeptide 8
Gene Aliases
CGB, hCGB
Chromosome Location
Chr.19: 49026385 - 49027549 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032928.1 NP_001028100.1 - 184 94
XM_015124374.2 XP_014979860.2 - 600 94
GenBank mRNA AY011015.1 - - 184 94

Gene Symbol
LOC100429320
Entrez Gene ID
100429320
Gene Name
choriogonadotropin subunit beta
Gene Aliases
-
Chromosome Location
Chr.19: 49007894 - 49009458 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028839671.1 XP_028695504.1 - 94

Gene Symbol
LOC114674119
Entrez Gene ID
114674119
Gene Name
choriogonadotropin subunit beta
Gene Aliases
-
Chromosome Location
Chr.19: 49015845 - 49017329 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028839670.1 XP_028695503.1 - 94

View Full Product Details