Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFRP4
Entrez Gene ID
574393
Gene Name
secreted frizzled related protein 4
Gene Aliases
-
Chromosome Location
Chr.3: 77251894 - 77261487 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032962.1 NP_001028134.1 - 632 76
GenBank mRNA AY321585.1 - - 632 76
JU473796.1 - - 76
JV043495.1 - - 76
JV635753.1 - - 76

View Full Product Details