Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL12B
Entrez Gene ID
694747
Gene Name
interleukin 12B
Gene Aliases
-
Chromosome Location
Chr.6: 156734270 - 156749918 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044725.1 NP_001038190.1 - 1003 79
GenBank mRNA U19841.1 - - 1003 79

View Full Product Details