Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTD1A
Entrez Gene ID
574122
Gene Name
demidefensin 2
Gene Aliases
RTD-3
Chromosome Location
Chr.8: 7715023 - 7717554 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF184157.1 - - 187 131

View Full Product Details