Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAMP3
Entrez Gene ID
707972
Gene Name
vesicle associated membrane protein 3
Gene Aliases
-
Chromosome Location
Chr.1: 216735891 - 216745801 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261368.1 NP_001248297.1 - 353 83
GenBank mRNA JU322518.1 - - 315 83
JU473306.1 - - 379 83
JV045674.1 - - 353 83
JV601919.1 - - 397 83

View Full Product Details