Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THOC3
Entrez Gene ID
695561
Gene Name
THO complex 3
Gene Aliases
EGK_17157
Chromosome Location
Chr.6: 173390474 - 173399197 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257489.1 NP_001244418.1 - 662 69
XM_015141442.2 XP_014996928.1 - 939 69
GenBank mRNA JU320930.1 - - 662 69
JU471220.1 - - 704 69
JV044030.1 - - 656 69

View Full Product Details