The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261171.1 | NP_001248100.1 | - | 505 | 125 |
GenBank mRNA | JU322857.1 | - | - | 505 | 125 |