Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYB5B
Entrez Gene ID
705093
Gene Name
cytochrome b5 type B
Gene Aliases
-
Chromosome Location
Chr.20: 56682502 - 56722230 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194167.3 NP_001181096.1 - 522 100
GenBank mRNA JU336517.1 - - 441 100
JU336518.1 - - 441 100
JU477327.1 - - 439 100
JV594712.1 - - 439 100
JV662348.1 - - 449 100
JV724166.1 - - 439 100

View Full Product Details