Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H9orf72
Entrez Gene ID
706828
Gene Name
chromosome 15 C9orf72 homolog
Gene Aliases
C9orf72
Chromosome Location
Chr.15: 56181632 - 56209219 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257526.1 NP_001244455.1 - 673 152
XM_015117390.2 XP_014972876.1 - 793 152
XM_015117391.2 XP_014972877.1 - 719 152
XM_028834299.1 XP_028690132.1 - 678 152
XM_028834301.1 XP_028690134.1 - 719 152
GenBank mRNA JU333327.1 - - 673 152
JU474639.1 - - 670 152
JV637336.1 - - 718 152

View Full Product Details