Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMRK1
Entrez Gene ID
703868
Gene Name
nicotinamide riboside kinase 1
Gene Aliases
C15H9orf95
Chromosome Location
Chr.15: 90493972 - 90522406 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015117794.2 XP_014973280.1 - 650 103
XM_028834280.1 XP_028690113.1 - 385 103
GenBank mRNA JU323108.1 - - 631 103

View Full Product Details