Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METAP2
Entrez Gene ID
712400
Gene Name
methionyl aminopeptidase 2
Gene Aliases
-
Chromosome Location
Chr.11: 95281073 - 95324672 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266460.1 NP_001253389.1 - 497 103
XM_015152393.2 XP_015007879.1 - 1077 103
GenBank mRNA JU321797.1 - - 513 103
JU474104.1 - - 487 103
JV043585.1 - - 497 103

View Full Product Details